Merck
  • Addition reaction of dialkyl disulfides to terminal alkynes catalyzed by a rhodium complex and trifluoromethanesulfonic acid.
Addition reaction of dialkyl disulfides to terminal alkynes catalyzed by a rhodium complex and trifluoromethanesulfonic acid.
Organic letters (2001-03-22)
M Arisawa, M Yamaguchi
ABSTRACT
[reaction: see text]. Addition of dialkyl disulfides to terminal alkynes is catalyzed by a rhodium-phosphine complex and trifluoromethanesulfonic acid giving (Z)-bis(alkylthio)olefins stereoselectively.
Sigma-Aldrich
Trifluoromethanesulfonic acid, ReagentPlus®, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Trifluoromethanesulfonic acid, reagent grade, 98%
Pricing and availability is not currently available.