- Synthesis of N-hydroxythiourea.
Synthesis of N-hydroxythiourea.
Journal of medicinal chemistry (1976-02-01)
M Sato, C H Stammer
PMID1249817
ABSTRACT
The synthesis of the title compound (1) was accomplished by the conversion of 2,4-dimethoxybenzylamine (2) into an isothiocyanate (3) using thiocarbonyl diimidazole. Treatment of 3 with hydroxylamine and removal of the DMB group with trifluoroacetic acid gave 1. N-Hydroxythiourea (1) showed no activity in the L1210 mouse tumor.