Merck
  • A thermostable L-aminoacylase from Thermococcus litoralis: cloning, overexpression, characterization, and applications in biotransformations.
A thermostable L-aminoacylase from Thermococcus litoralis: cloning, overexpression, characterization, and applications in biotransformations.
Extremophiles : life under extreme conditions (2002-05-16)
Helen S Toogood, Edward J Hollingsworth, Rob C Brown, Ian N Taylor, Stephen J C Taylor, Ray McCague, Jennifer A Littlechild
ABSTRACT
A thermostable L-aminoacylase from Thermococcus litoralis was cloned, sequenced, and overexpressed in Escherichia coli. The enzyme is a homotetramer of 43 kDa monomers and has an 82% sequence identity to an aminoacylase from Pyrococcus horikoshii and 45% sequence identity to a carboxypeptidase from Sulfolobus solfataricus. It contains one cysteine residue that is highly conserved among aminoacylases. Cell-free extracts of the recombinant enzyme were characterized and were found to have optimal activity at 85 degrees C in Tris-HCl at pH 8.0. The recombinant enzyme is thermostable, with a half-life of 25 h at 70 degrees C. Aminoacylase inhibitors, such as mono-tert-butyl malonate, had only a slight effect on activity. The enzyme was partially inhibited by EDTA and p-hydroxymercuribenzoate, suggesting that the cysteine residue and a metal ion are important, but not essential, for activity. Addition of Zn2+ and Co2+ to the apoenzyme increased the enzyme activity, whereas Sn4+ and Cu2+ almost completely abolished enzyme activity. The enzyme was most specific for substrates containing N-benzoyl- or N-chloroacetyl-amino acids. preferring substrates containing hydrophobic, uncharged, or weakly charged amino acids such as phenylalanine, methionine, and cysteine.
Sigma-Aldrich
mono-tert-Butyl malonate, 95%
Pricing and availability is not currently available.