Merck
  • Thermodynamics of the interaction between oxytocin and its myometrial receptor in sheep: a stepwise binding mechanism.
Thermodynamics of the interaction between oxytocin and its myometrial receptor in sheep: a stepwise binding mechanism.
Biochemical pharmacology (2014-07-11)
Vladimir Pliska, Gerd Folkers, Vojtěch Spiwok
ABSTRACT
Entropy (ΔS), enthalpy (ΔH) and heat capacity (ΔCp) changes attending the oxytocin interaction with its two binding sites on myometrial cell membranes in sheep were derived from the temperature dependence of Kd values. The high affinity oxytocin site (Kd on the order of 10(-9)mol l(-1), 25 °C), ascribed to the oxytocin receptor (OXTR), is entropy-driven in the temperature range 0-37 °C. Enthalpy component prevails as a driving force in the binding to the low affinity site (Kd ≈ 10(-7)) within the higher temperature range. ΔCp values in both cases do not differ significantly from zero but become highly relevant in the presence of a GTP analog (10(-4)M GTP-γS). Under these conditions, ΔCp in the low site interaction becomes negative and ΔS is shifted toward negative values (enthalpy drift); ΔCp of the high affinity site rises to a high positive value and the interaction is even more strongly entropy driven. Atosiban, a competitive antagonist of oxytocin at OXTR displays a single significant binding site on myometrial cells (Kd about 10(-7)mol l(-1)). Thermodynamic profiles of atosiban and the low affinity oxytocin site show conspicuous similarities, indicating that the inhibitor is bound to the low affinity site, and not, with a lower affinity, to the putative receptor protein. It is suggested that the interaction of oxytocin with its responding system on myometrial membranes follows in two distinct steps that are likely to be associated with several independent binding domains in the GPCR receptor.
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Trypsin inhibitor from Glycine max (soybean), lyophilized powder
Sigma-Aldrich
Bacitracin, from Bacillus licheniformis, ≥65 IU/mg
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Bacitracin, from Bacillus licheniformis, ≥65 IU/mg
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Benzamidine, ≥95.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Oxytocin, lyophilized powder, ~15 IU/mg solid (Prepared from synthetic oxytocin)
Sigma-Aldrich
Oxytocin
Sigma-Aldrich
Benzamidine hydrochloride hydrate, ≥99%
USP
Oxytocin, United States Pharmacopeia (USP) Reference Standard
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Oxytocin, European Pharmacopoeia (EP) Reference Standard