Merck
  • A designed supramolecular protein assembly with in vivo enzymatic activity.
A designed supramolecular protein assembly with in vivo enzymatic activity.
Science (New York, N.Y.) (2014-12-20)
Woon Ju Song, F Akif Tezcan
ABSTRACT
The generation of new enzymatic activities has mainly relied on repurposing the interiors of preexisting protein folds because of the challenge in designing functional, three-dimensional protein structures from first principles. Here we report an artificial metallo-β-lactamase, constructed via the self-assembly of a structurally and functionally unrelated, monomeric redox protein into a tetrameric assembly that possesses catalytic zinc sites in its interfaces. The designed metallo-β-lactamase is functional in the Escherichia coli periplasm and enables the bacteria to survive treatment with ampicillin. In vivo screening of libraries has yielded a variant that displays a catalytic proficiency [(k(cat)/K(m))/k(uncat)] for ampicillin hydrolysis of 2.3 × 10(6) and features the emergence of a highly mobile loop near the active site, a key component of natural β-lactamases to enable substrate interactions.
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Ampicillin trihydrate, 900-1050 μg/mg anhydrous basis (HPLC)
Supelco
Ampicillin trihydrate, VETRANAL®, analytical standard
Supelco
Ampicillin Trihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Zinc, foil, 15mm disks, thickness 0.125mm, as rolled, 99.95+%
Zinc, foil, 10mm disks, thickness 0.25mm, as rolled, 99.99+%
Zinc, foil, 25mm disks, thickness 0.05mm, as rolled, 99.95+%
Zinc, foil, 1m coil, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, 0.3m coil, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, 15mm disks, thickness 0.005mm, 99.9%
Zinc, foil, 1m coil, thickness 0.1mm, as rolled, 99.95+%
Zinc, foil, 25mm disks, thickness 0.175mm, as rolled, 99.95+%
Zinc, wire reel, 2m, diameter 1.0mm, 99.99+%
Zinc, wire reel, 2m, diameter 1.0mm, 99.999%
Zinc, wire reel, 10m, diameter 0.5mm, as drawn, 99.99+%
Zinc, wire reel, 5m, diameter 3.0mm, extruded, 99.9%
Zinc, wire reel, 5m, diameter 0.025mm, as drawn, 99%
Zinc, wire reel, 10m, diameter 0.025mm, as drawn, 99%
Zinc, wire reel, 1m, diameter 0.125mm, as drawn, 99%
Zinc, wire reel, 100m, diameter 1.0mm, extruded, 99.9%