Merck
  • Genetic hemoglobin disorders rather than iron deficiency are a major predictor of hemoglobin concentration in women of reproductive age in rural prey Veng, Cambodia.
Genetic hemoglobin disorders rather than iron deficiency are a major predictor of hemoglobin concentration in women of reproductive age in rural prey Veng, Cambodia.
The Journal of nutrition (2014-12-21)
Crystal D Karakochuk, Kyly C Whitfield, Susan I Barr, Yvonne Lamers, Angela M Devlin, Suzanne M Vercauteren, Hou Kroeun, Aminuzzaman Talukder, Judy McLean, Timothy J Green
ABSTRACT
Anemia is common in Cambodian women. Potential causes include micronutrient deficiencies, genetic hemoglobin disorders, inflammation, and disease. We aimed to investigate factors associated with anemia (low hemoglobin concentration) in rural Cambodian women (18-45 y) and to investigate the relations between hemoglobin disorders and other iron biomarkers. Blood samples were obtained from 450 women. A complete blood count was conducted, and serum and plasma were analyzed for ferritin, soluble transferrin receptor (sTfR), folate, vitamin B-12, retinol binding protein (RBP), C-reactive protein (CRP), and α1 acid glycoprotein (AGP). Hemoglobin electrophoresis and multiplex polymerase chain reaction were used to determine the prevalence and type of genetic hemoglobin disorders. Overall, 54% of women had a genetic hemoglobin disorder, which included 25 different genotypes (most commonly, hemoglobin E variants and α(3.7)-thalassemia). Of the 420 nonpregnant women, 29.5% had anemia (hemoglobin <120 g/L), 2% had depleted iron stores (ferritin <15 μg/L), 19% had tissue iron deficiency (sTfR >8.3 mg/L), <3% had folate deficiency (<3 μg/L), and 1% had vitamin B-12 deficiency (<150 pmol/L). Prevalences of iron deficiency anemia (IDA) were 14.2% and 1.5% in those with and without hemoglobin disorders, respectively. There was no biochemical evidence of vitamin A deficiency (RBP <0.7 μmol/L). Acute and chronic inflammation were prevalent among 8% (CRP >5 mg/L) and 26% (AGP >1 g/L) of nonpregnant women, respectively. By using an adjusted linear regression model, the strongest predictors of hemoglobin concentration were hemoglobin E homozygous disorder and pregnancy status. Other predictors were 2 other heterozygous traits (hemoglobin E and Constant Spring), parity, RBP, log ferritin, and vitamin B-12. Multiple biomarkers for anemia and iron deficiency were significantly influenced by the presence of hemoglobin disorders, hence reducing their diagnostic sensitivity. Further investigation of the unexpectedly low prevalence of IDA in Cambodian women is warranted.
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Ferritin from human spleen, Type V, 10 μg/mL in 0.15 M NaCl, 10 mM Tris, pH 8.0, containing 0.1% sodium azide
Sigma-Aldrich
Ferritin from equine spleen, Type I, saline solution
Iron, wire reel, 5m, diameter 0.025mm, as drawn, 99.99+%
Iron, wire reel, 1m, diameter 0.5mm, as drawn, 99.998%
Iron, wire reel, 0.5m, diameter 0.25mm, hard, 99.99+%
Iron, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.99+%
Iron, wire reel, 5m, diameter 0.25mm, hard, 99.99+%
Iron, wire reel, 50m, diameter 0.5mm, as drawn, 99.95%
Iron, wire reel, 2m, diameter 0.25mm, annealed, 99.99+%
Iron, wire reel, 0.05m, diameter 0.5mm, as drawn, 99.998%
Iron, rod, 500mm, diameter 12.7mm, as drawn, 99.95%
Iron, foil, 50x50mm, thickness 2mm, as rolled, 99.99+%
Iron, tube, 200mm, outside diameter 4.0mm, inside diameter 3.6mm, wall thickness 0.2mm, as drawn, 99.5%
Iron, rod, 200mm, diameter 25mm, as drawn, 98+%
Iron, tube, 100mm, outside diameter 8.0mm, inside diameter 5mm, wall thickness 1.5mm, annealed, 99.5%
Iron, foil, 300x300mm, thickness 1.0mm, as rolled, 99.5%
Iron, foil, 300x300mm, thickness 2.0mm, as rolled, armco« soft ingot 99.8%
Iron, wire reel, 1m, diameter 0.025mm, as drawn, 99.99+%
Iron, foil, 300x300mm, thickness 0.15mm, hard, 99.5%
Iron, foil, 50x50mm, thickness 0.25mm, as rolled, 99.99+%
Iron, wire reel, 1000m, diameter 0.125mm, hard, 99.5%
Iron, foil, 10mm disks, thickness 0.005mm, 99.85%
Iron, tube, 500mm, outside diameter 3.18mm, inside diameter 1.96mm, wall thickness 0.61mm, as drawn, 99.5%
Iron, tube, 200mm, outside diameter 5.0mm, inside diameter 4.5mm, wall thickness 0.25mm, as drawn, 99.5%
Iron, wire reel, 20m, diameter 0.025mm, as drawn, 99.99+%
Iron, wire reel, 1000m, diameter 0.25mm, hard, 99.5%