Merck
  • Earth history. U-Pb geochronology of the Deccan Traps and relation to the end-Cretaceous mass extinction.
Earth history. U-Pb geochronology of the Deccan Traps and relation to the end-Cretaceous mass extinction.
Science (New York, N.Y.) (2014-12-17)
Blair Schoene, Kyle M Samperton, Michael P Eddy, Gerta Keller, Thierry Adatte, Samuel A Bowring, Syed F R Khadri, Brian Gertsch
ABSTRACT
The Chicxulub asteroid impact (Mexico) and the eruption of the massive Deccan volcanic province (India) are two proposed causes of the end-Cretaceous mass extinction, which includes the demise of nonavian dinosaurs. Despite widespread acceptance of the impact hypothesis, the lack of a high-resolution eruption timeline for the Deccan basalts has prevented full assessment of their relationship to the mass extinction. Here we apply uranium-lead (U-Pb) zircon geochronology to Deccan rocks and show that the main phase of eruptions initiated ~250,000 years before the Cretaceous-Paleogene boundary and that >1.1 million cubic kilometers of basalt erupted in ~750,000 years. Our results are consistent with the hypothesis that the Deccan Traps contributed to the latest Cretaceous environmental change and biologic turnover that culminated in the marine and terrestrial mass extinctions.
Sigma-Aldrich
Zirconium, powder, −100 mesh
Sigma-Aldrich
Zirconium, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Lead, powder, −100 mesh, 99.95% trace metals basis
Sigma-Aldrich
Zirconium, rod, diam. 6.35 mm, ≥99% trace metals basis
Sigma-Aldrich
Zirconium, sponge, ≥99% trace metals basis
Sigma-Aldrich
Lead, shot, <2 mm, 99.9% trace metals basis
Sigma-Aldrich
Lead, shot, 1-3 mm, 99.995% trace metals basis
Zirconium, foil, 15mm disks, thickness 0.01mm, 99.2%
Zirconium, foil, 15mm disks, thickness 0.025mm, annealed, 99.2%
Zirconium, foil, 25mm disks, thickness 0.009mm, 99.2%
Zirconium, foil, 300x300mm, thickness 0.25mm, annealed, 99.2%
Zirconium, foil, 25mm disks, thickness 0.075mm, annealed, 99.2%
Zirconium, foil, 50x50mm, thickness 0.25mm, annealed, 99.2%
Zirconium, foil, 10mm disks, thickness 0.016mm, 99.2%
Zirconium, foil, 1m coil, thickness 0.016mm, 99.2%
Zirconium, foil, 0.5m coil, thickness 0.075mm, annealed, 99.2%
Zirconium, foil, 25x25mm, thickness 0.5mm, annealed, 99.2%
Zirconium, foil, 0.5m coil, thickness 0.125mm, annealed, 99.2%
Zirconium, foil, 25mm disks, thickness 0.016mm, 99.2%
Zirconium, foil, 15mm disks, thickness 0.02mm, 99.2%
Zirconium, foil, 0.5m coil, thickness 0.025mm, annealed, 99.2%
Zirconium, foil, 10mm disks, thickness 0.25mm, annealed, 99.2%
Zirconium, foil, 10mm disks, thickness 0.5mm, annealed, 99.2%
Zirconium, foil, 10mm disks, thickness 0.01mm, 99.2%
Zirconium, foil, 15mm disks, thickness 0.05mm, annealed, 99.2%
Zirconium, foil, 25mm disks, thickness 0.01mm, 99.2%
Zirconium, foil, 1m coil, thickness 0.025mm, annealed, 99.2%
Zirconium, foil, 25mm disks, thickness 0.025mm, annealed, 99.2%
Zirconium, foil, 10mm disks, thickness 0.01mm, 99.2%
Zirconium, foil, 10mm disks, thickness 0.025mm, annealed, 99.2%