Merck
  • Skin extracellular matrix stimulation following injection of a hyaluronic acid-based dermal filler in a rat model.
Skin extracellular matrix stimulation following injection of a hyaluronic acid-based dermal filler in a rat model.
Plastic and reconstructive surgery (2014-11-22)
Sumit Paliwal, Steven Fagien, Xiaojian Sun, Tyler Holt, Thomas Kim, Christopher K Hee, Dennis Van Epps, Darin J Messina
ABSTRACT
Hyaluronic acid-based dermal fillers have gained rapid acceptance for treating facial wrinkles and deep tissue folds. Although their space-filling properties are well understood, this study evaluates the cellular and molecular changes in skin, as a secondary effect, following injection of a commercially available, 24-mg/ml, cross-linked hyaluronic acid-based filler (HYC-24L+) in a rodent model. Sprague-Dawley rats, aged 2 to 4 months, were injected intradermally with 20 μl of HYC-24L+ using a linear threading technique and followed to 12 weeks after injection. Untreated skin and saline injection were used as study controls. Enzyme-linked immunosorbent assay and reverse-transcriptase polymerase chain reaction methods were used to investigate changes in the expression of several extracellular matrix proteins and genes over time. HYC-24L+ significantly increased the protein expression levels of collagen types I and III in rat dermal tissue for up to 12 weeks. The ratio of collagen type III to type I protein, however, remained unchanged, suggesting maintenance of collagen homeostasis. A significant increase in dermal elastin after HYC-24L+ injection was also observed. Gene expression analysis confirmed that several genes associated with extracellular matrix production and assembly were also transiently up-regulated, and that these changes temporally preceded those observed at the protein level. In addition to its well-understood space-filling function, as a secondary effect, the authors demonstrate that HYC-24L+ stimulates the production of several extracellular matrix components, including dermal collagen and elastin.
Sigma-Aldrich
Elastin, soluble from bovine neck ligament, salt-free, lyophilized powder
Sigma-Aldrich
Hyaluronic acid sodium salt from bovine vitreous humor
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 150,000-300,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,750,000-2,000,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 500,000-750,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 90,000-110,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 30,000-50,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,200
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,200,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 70,000-120,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 50,000-70,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 15,000-30,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 8,000-15,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,500,000-1,750,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,000,000-1,250,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 2,000,000-2,400,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 300,000-500,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 10,000-30,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 120,000-350,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus zooepidemicus, bacterial glycosaminoglycan polysaccharide
Sigma-Aldrich
Hyaluronic acid sodium salt from rooster comb, avian glycosaminoglycan polysaccharide
Sigma-Aldrich
Elastin from bovine neck ligament, powder
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 130,000-150,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 750,000-1,000,000
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,250,000-1,500,000