Merck
  • Uncemented femoral revision arthroplasty using a modular tapered, fluted titanium stem: 5- to 16-year results of 163 cases.
Uncemented femoral revision arthroplasty using a modular tapered, fluted titanium stem: 5- to 16-year results of 163 cases.
Acta orthopaedica (2014-09-02)
Dieter C Wirtz, Sascha Gravius, Rudolf Ascherl, Miguel Thorweihe, Raimund Forst, Ulrich Noeth, Uwe M Maus, Matthias D Wimmer, Günther Zeiler, Moritz C Deml
ABSTRACT
Due to the relative lack of reports on the medium- to long-term clinical and radiographic results of modular femoral cementless revision, we conducted this study to evaluate the medium- to long-term results of uncemented femoral stem revisions using the modular MRP-TITAN stem with distal diaphyseal fixation in a consecutive patient series. We retrospectively analyzed 163 femoral stem revisions performed between 1993 and 2001 with a mean follow-up of 10 (5-16) years. Clinical assessment included the Harris hip score (HHS) with reference to comorbidities and femoral defect sizes classified by Charnley and Paprosky. Intraoperative and postoperative complications were analyzed and the failure rate of the MRP stem for any reason was examined. Mean HHS improved up to the last follow-up (37 (SD 24) vs. 79 (SD 19); p<0.001). 99 cases (61%) had extensive bone defects (Paprosky IIB-III). Radiographic evaluation showed stable stem anchorage in 151 cases (93%) at the last follow-up. 10 implants (6%) failed for various reasons. Neither a breakage of a stem nor loosening of the morse taper junction was recorded. Kaplan-Meier survival analysis revealed a 10-year survival probability of 97% (95% CI: 95-100). This is one of the largest medium- to long-term analyses of cementless modular revision stems with distal diaphyseal anchorage. The modular MRP-TITAN was reliable, with a Kaplan-Meier survival probability of 97% at 10 years.
Sigma-Aldrich
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Sigma-Aldrich
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Titanium, IRMM®, certified reference material, 0.5 mm foil
Titanium, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Titanium, rod, 200mm, diameter 25mm, annealed, 99.6+%
Titanium, tube, 200mm, outside diameter 2.03mm, inside diameter 1.55mm, wall thickness 0.24mm, annealed, 99.6+%
Titanium, rod, 200mm, diameter 5mm, annealed, 99.6+%
Titanium, rod, 1000mm, diameter 3.0mm, annealed, 99.6+%
Titanium, wire reel, 1m, diameter 0.20mm, annealed, 99.6+%
Titanium, wire reel, 1m, diameter 0.5mm, as drawn, 99.6+%
Titanium, wire reel, 100m, diameter 0.25mm, annealed, 99.6+%
Titanium, wire reel, 20m, diameter 1.5mm, as drawn, 99.6+%
Titanium, wire reel, 1m, diameter 1.5mm, as drawn, 99.6+%
Titanium, wire reel, 20m, diameter 0.05mm, hard, 99.8%
Titanium, wire reel, 2m, diameter 1.0mm, as drawn, 99.99+%
Titanium, wire reel, 200m, diameter 0.125mm, annealed, 99.6+%
Titanium, wire reel, 5m, diameter 0.20mm, annealed, 99.6+%
Titanium, wire reel, 25m, diameter 0.20mm, annealed, 99.6+%
Titanium, wire reel, 500m, diameter 0.20mm, annealed, 99.6+%
Titanium, wire reel, 200m, diameter 0.20mm, annealed, 99.6+%
Titanium, wire reel, 20m, diameter 0.125mm, annealed, 99.6+%
Titanium, wire reel, 5m, diameter 0.25mm, as drawn, 99.99+%
Titanium, wire reel, 25m, diameter 0.25mm, annealed, 99.6+%
Titanium, wire reel, 200m, diameter 0.25mm, as drawn, 99.6+%
Titanium, wire reel, 25m, diameter 0.25mm, as drawn, 99.6+%