Merck
  • Strength and function response to clinical interventions of older women categorized by weakness and low lean mass using classifications from the Foundation for the National Institute of Health sarcopenia project.
Strength and function response to clinical interventions of older women categorized by weakness and low lean mass using classifications from the Foundation for the National Institute of Health sarcopenia project.
The journals of gerontology. Series A, Biological sciences and medical sciences (2014-08-20)
Maren S Fragala, Thuy-Tien L Dam, Vanessa Barber, James O Judge, Stephanie A Studenski, Peggy M Cawthon, Robert R McLean, Tamara B Harris, Luigi Ferrucci, Jack M Guralnik, Douglas P Kiel, Stephen B Kritchevsky, Michelle D Shardell, Maria T Vassileva, Anne M Kenny
ABSTRACT
The Foundation for the National Institutes of Health Sarcopenia Project developed data-driven cut-points for clinically meaningful weakness and low lean body mass. This analysis describes strength and function response to interventions based on these classifications. In data from four intervention studies, 378 postmenopausal women with baseline and 6-month data were evaluated for change in grip strength, appendicular lean mass corrected for body mass index, leg strength and power, and short physical performance battery (SPPB). Clinical interventions included hormones, exercise, and nutritional supplementation. Differences in outcomes were evaluated between (i) those with and without weakness and (ii) those with weakness and low lean mass or with one but not the other. We stratified analyses by slowness (walking speed ≤ 0.8 m/s) and by treatment assignment. The women (72±7 years; body mass index of 26±5kg/m(2)) were weak (33%), had low lean mass (14%), or both (6%). Those with weakness increased grip strength, lost less leg power, and gained SPPB score (p < .05) compared with nonweak participants. Stratified analyses were similar for grip strength and SPPB. With lean mass in the analysis, individuals with weakness had larger gains in grip strength and SPPB scores regardless of low lean mass (p < .01). Older women with clinically meaningful muscle weakness increased grip strength and SPPB, regardless of the presence of low lean mass following treatment with interventions for frailty. Thus, results suggest that muscle weakness, as defined by the Foundation for the National Institutes of Health Sarcopenia Project, appears to be a treatable symptom.
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
Calcium citrate tetrahydrate, 99%
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
Estradiol, meets USP testing specifications
Estradiol hemihydrate, European Pharmacopoeia (EP) Reference Standard
Supelco
17β-Estradiol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material