Merck
  • Comparison of three gamma-turn mimetic scaffolds incorporated into angiotensin II.
Comparison of three gamma-turn mimetic scaffolds incorporated into angiotensin II.
Bioorganic & medicinal chemistry (2000-10-12)
S Lindman, G Lindeberg, F Nyberg, A Karlén, A Hallberg
ABSTRACT
Rigidification of peptides by cyclization and iterative incorporation of well-defined secondary structure mimetics constitutes one approach to the design of non-peptidergic structures with better defined conformations. We herein present the synthesis of a potential gamma-turn mimetic scaffold, and its incorporation in the 3-5 position of angiotensin II. Two analogues of angiotensin II (Ang II) incorporating this 1,3,5-trisubstituted benzene gamma-turn scaffold were synthesized. Evaluation of the compounds in a radioligand binding assay showed that they lacked affinity to the AT1 receptor. To rationalize these results a geometrical and electrostatical comparison with Ang II analogues encompassing a bicyclic scaffold that delivered inactive pseudo peptides and an azepine scaffold producing highly active ligands was made. This analysis did not provide a clear rationale for the inactivity of the benzene gamma-turn scaffolds.
Sigma-Aldrich
3-Bromo-5-iodobenzoic acid, 97%
Pricing and availability is not currently available.