Merck
  • MIB-1 proliferation index predicts survival among patients with grade II astrocytoma.
MIB-1 proliferation index predicts survival among patients with grade II astrocytoma.
Journal of neuropathology and experimental neurology (1998-10-24)
P E McKeever, M S Strawderman, B Yamini, A A Mikhail, M Blaivas
ABSTRACT
The purpose of this study was to determine whether a relationship existed between MIB-1 labeling index (LI) percentages and survival in patients with grade II astrocytomas. From archival paraffin-embedded surgical specimens of 50 patients of the University of Michigan Medical Center with World Health Organization grade II astrocytomas, 22 patients had a Ki-67 LI of less than or equal to 2.0; and 28 patients had a MIB-1 LI of more than 2.0. Over a median follow-up interval of 10 years, ranging up to 16 years, 23% (n = 5) died of tumor in the first group while 82% (n = 23) died in the second group, a distinct difference in survival between these groups. Univariate analysis showed that a high MIB-1 predicted shorter survival (p < 0.0001), and that increased age was associated with shorter survival (p = 0.007). Gender, tumor location and radiotherapy had no significant association with survival. When adjusting for these (excluding tumor location) in the Cox proportional hazards model simultaneously, MIB-1 and age were independently prognostic. The hazard ratios were 1.301 per 1% MIB-1 LI (p = 0.0001), and 1.045 per year of age (p = 0.0028). From other studies, we know that histopathologic grade and age predict survival for glioma patients. However, even within grade II astrocytomas there is still a wide heterogeneity in how long a patient survives. We conclude that among grade II astrocytomas older patients and, independently, patients with higher MIB-1 labeling index have shorter survival.