- The human homolog of Drosophila cornichon protein is differentially expressed in alloactivated T-cells.
The human homolog of Drosophila cornichon protein is differentially expressed in alloactivated T-cells.
To identify novel genes induced in the early stage of T-cell activation, mRNA expression in alloactivated human lymphocytes was examined. Differential display-reverse transcription PCR analysis revealed a 207-bp cDNA fragment which was upregulated 24 h after allostimulation of a human T-cell line. The corresponding complete 1396 bp cDNA, named TGAM77, encodes a predicted 134 amino acid protein which shares 63% homology with the cornichon (cni) protein of Drosophila melanogaster. Upregulation of TGAM77 mRNA in the early phase of T-cell activation was confirmed by Northern blot and RT-PCR analysis of activated human lymphocytes. TGAM77 mRNA is expressed in a variety of human tissues with various expression levels. In analogy to cni which is involved in an epidermal growth factor-like signaling pathway inducing cellular asymmetry in Drosophila oogenesis, TGAM77 might function in similar signaling establishing vectorial re-localization and concentration of signaling events in T-cell activation.