Merck
  • Enantioselective gas chromatographic analysis of aqueous samples by on-line derivatisation. Application to enzymatic reactions.
Enantioselective gas chromatographic analysis of aqueous samples by on-line derivatisation. Application to enzymatic reactions.
Journal of chromatography. A (2008-02-02)
John H M Mommers, Stefaan M A de Wildeman, Wilma A F Koolen, Alexander L L Duchateau
ABSTRACT
A new gas chromatography (GC) method is presented for analysing both the conversion and the enantiomeric excess (e.e.) of samples from alcohol dehydrogenase reactions. The chiral compounds studied were a series of saturated, straight chain alcohols, ranging from 2-butanol to 2-heptanol. The alcohols were converted to the corresponding trifluoroacetylated derivatives by injecting trifluoroacetic anhydride onto the column shortly after injection of the aqueous samples in split-injection mode (1:100) onto a Chiraldex G-TA capillary GC column. Injecting seven hundred aqueous enzymatic reaction mixtures according to the above-mentioned procedure revealed no noticeable loss of column performance. Using the new GC method, conventional sample work-up procedures such as extraction and off-line derivatisation are eliminated and throughput of samples is significantly enhanced.
Product Description
Supelco
Astec® CHIRALDEX G-TA Capillary GC Column, L × I.D. 20 m × 0.25 mm, df 0.12 μm
Supelco
Astec® CHIRALDEX G-TA Capillary GC Column, L × I.D. 50 m × 0.25 mm, df 0.12 μm
Supelco
Astec® CHIRALDEX GC Column Screening Kit, L × I.D. 30 m × 0.25 mm, pkg of 1 kit
Supelco
Astec® CHIRALDEX G-TA Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.12 μm