Merck
  • O-Monoacyltartaric acid catalyzed enantioselective conjugate addition of a boronic acid to dienones: application to the synthesis of optically active cyclopentenones.
O-Monoacyltartaric acid catalyzed enantioselective conjugate addition of a boronic acid to dienones: application to the synthesis of optically active cyclopentenones.
Organic letters (2014-09-24)
Masaharu Sugiura, Ryo Kinoshita, Makoto Nakajima
ABSTRACT
Enantioselective conjugate addition of styrylboronic acid to dienones was effectively catalyzed by an O-monoacyltartaric acid to afford monostyrylated products with good enantioselectivity. The RCM of the monostyrylated products using the Hoveyda-Grubbs II catalyst afforded optically active cyclopentenones, including a synthetic intermediate of the antitumor agent TEI-9826. The study shows that a diene additive such as 1,6-heptadiene or diallyl ether was essential for the RCM.
Product Description
Supelco
Tartaric Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
L-(+)-Tartaric acid, ≥99.5%
Pricing and availability is not currently available.
Supelco
L-(+)-Tartaric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Sigma-Aldrich
L-(+)-Tartaric acid, puriss. p.a., reag. ISO, 99.5-101.0% (calc. to the dried substance)
Pricing and availability is not currently available.
Sigma-Aldrich
L-(+)-Tartaric acid, puriss., meets analytical specification of Ph. Eur., NF, 99.7-100.5% (calc. to the dried substance), powder
Pricing and availability is not currently available.
Sigma-Aldrich
L-(+)-Tartaric acid, ≥99.7%, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
L-(+)-Tartaric acid, ACS reagent, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
L-(+)-Tartaric acid, BioXtra
Pricing and availability is not currently available.
Sigma-Aldrich
L-(+)-Tartaric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Cyclopenten-1-one, 98%
Pricing and availability is not currently available.