Merck
  • Intra-individual comparison of magnesium citrate and sodium phosphate for bowel preparation at CT colonography: automated volumetric analysis of residual fluid for quality assessment.
Intra-individual comparison of magnesium citrate and sodium phosphate for bowel preparation at CT colonography: automated volumetric analysis of residual fluid for quality assessment.
Clinical radiology (2014-09-23)
P Bannas, J Bakke, A Munoz del Rio, P J Pickhardt
ABSTRACT
To perform an objective, intra-individual comparison of residual colonic fluid volume and attenuation associated with the current front-line laxative magnesium citrate (MgC) versus the former front-line laxative sodium phosphate (NaP) at CT colonography (CTC). This retrospective Health Insurance and Portability and Accountability Act-compliant study had institutional review board approval; informed consent was waived. The study cohort included 250 asymptomatic adults (mean age at index 56.1 years; 124 male/126 female) who underwent CTC screening twice over a 5 year interval. Colon catharsis at initial and follow-up screening employed single-dose NaP and double-dose MgC, respectively, allowing for intra-patient comparison. Automated volumetric analysis of residual colonic fluid volume and attenuation was performed on all 500 CTC studies. Colonic fluid volume <200 ml and mean attenuation between 300-900 HU were considered optimal. Paired t-test and McNemar's test were used to compare differences. Residual fluid volumes <200 ml were recorded in 192 examinations (76.8%) following MgC and in 204 examinations (81.6%) following NaP (p = 0.23). The mean total residual fluid volume was 155 ± 114 ml for MgC and 143 ± 100 ml for NaP (p = 0.01). The attenuation range of 300-900 HU was significantly more frequent for MgC (n = 220, 88%) than for NaP (n = 127, 50.8%; p < 0.001). Mean fluid attenuation was significantly lower for MgC (700 ± 165 HU) than for NaP (878 ± 155 HU; p < 0.001). Concomitant presence of both optimal fluid volume and attenuation was significantly more frequent for MgC 65.2% than for NaP (38%; p < 0.001). Objective intra-individual comparison using automated volumetric analysis suggests that the replacement of NaP by MgC as the front-line laxative for CTC has not compromised overall examination quality.
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Sodium phosphate dibasic heptahydrate, meets USP testing specifications
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium phosphate dibasic heptahydrate, ≥99.99% trace metals basis
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sodium phosphate dibasic heptahydrate, ACS reagent, 98.0-102.0%
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%