Merck
  • The PSI family of nuclear proteins is required for growth in arabidopsis.
The PSI family of nuclear proteins is required for growth in arabidopsis.
Plant molecular biology (2014-07-27)
Nils Stührwohldt, Jens Hartmann, Renate I Dahlke, Claudia Oecking, Margret Sauter
ABSTRACT
PSI1 was identified as a gene that is co-expressed with the phytosulfokine (PSK) receptor genes PSKR1 and PSKR2 in Arabidopsis thaliana. It represents a plant-specific protein family of unknown function with six members in two clades. Clade 1 members PSI1, PSI2 and PSI3 were characterized in this study. All three are nuclear localized. A predicted N-terminal myristoylation site was functionally analyzed. psi1-1 seedlings have shorter roots and hypocotyls. This growth-retarded phenotype was restored by expression of either wildtype PSI1 or PSI1 G2A with a mutated myristate attachment site in the psi1-1 background suggesting that myristate attachment was not essential for PSI1 function. psi2-1 and psi3-1 seedlings have a wildtype phenotype but overexpression of PSI1 or PSI2 promoted seedling growth. PSI2 activity appears to be linked to PSK signaling as psi2-1 and psi2-1 psi3-1 roots are unresponsive to PSK. PSI3 functions in vegetative plant growth synergistic with PSI2. psi3-1 and particularly psi2-1 psi3-1 rosettes are small. Overexpression of PSI3 promoted plant growth indicating that PSI3 is limiting at the vegetative stage. Severe dwarfism of psi2-1 psi3-1 plants results from reduced cell growth and proliferation and premature leaf growth arrest. Plants further display reduced fertility and premature senescence revealing a crucial function of PSI proteins in vegetative growth and reproduction. Psi single and double knock-out plants have less and PSI3ox plants have more starch compared to wt and growth retardation is partially rescued by sucrose. Our studies reveal a crucial function of the nuclear-localized PSI proteins in growth possibly through metabolic control.
Sigma-Aldrich
Starch from potato, Soluble
Sigma-Aldrich
Starch from potato, suitable for electrophoresis
Sigma-Aldrich
Starch, from potato, tested according to Ph. Eur.
Sigma-Aldrich
Starch, puriss. p.a., from potato, reag. ISO, reag. Ph. Eur., soluble