Merck
Fate of phosphorus in diluted urine with tap water.
Chemosphere (2014-07-30)
Xiaoning Liu, Guoqi Wen, Huihui Wang, Xiaoqi Zhu, Zhengyi Hu
ABSTRACT
P loss during the fresh urine storage process is inevitable because of the presence of Ca and Mg. Dilution is one of the most important parameters influencing urine composition and subsequent P recovery. This study aimed to investigate the fate of P in urine with different dilution ratios (Vwater/Vurine, i.e., 0/100, 25/75, 50/50 and 75/25). The results indicate that the percentage of P loss increased from 43% to 76% as the dilution ratio increased from 0/100 to 75/25 because of more Ca and Mg obtained from tap water. Meanwhile, P removal efficiency through struvite precipitation decreased from 51% to 8% because of lower supersaturation ratio as a result of dilution. Struvite crystals with high purity were still obtained even under a dilution ratio of 50/50 urine solution. Batch experiments were also performed to study the influence of temperature (15-35°C) on P recovery and crystal size. For different dilution ratios of urine solutions, no significant discrepancy for the P removal efficiencies were observed at 15 and 35°C, whereas the P removal efficiencies at 25°C showed an increasing gap with those at 15 and 35°C. The largest average crystal sizes were found at 25°C, which was opposite to the trend of P removal efficiency.
Product Description
Supelco
Water, for TOC analysis
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Water, ACS reagent, for ultratrace analysis
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Magnesium, chips, 6-35 mesh, 99.98% trace metals basis
Sigma-Aldrich
Magnesium, ReagentPlus®, ribbon, ≥99% trace metals basis
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Magnesium, turnings, reagent grade, 98%
Sigma-Aldrich
Magnesium, turnings, 5-25 mm, 99.95% trace metals basis
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Magnesium, ribbon, ≥99.0% Mg basis
Magnesium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 1.1mm, as drawn, 99.9+%
Magnesium, microfoil, 50x50mm, thinness 1.0μm, specific density 174μg/cm2, 6 micron aluminum permanent support, 99.9%
Magnesium, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.9+%
Magnesium, wire reel, 2m, diameter 1.1mm, as drawn, 99.9+%
Magnesium, wire reel, 1m, diameter 0.5mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 0.25mm, as drawn, 99.9+%
Magnesium, microleaf, 25x25mm, thinness 1.0μm, specific density 174μg/cm2, removable support, 99.9%
Magnesium, wire reel, 2m, diameter 0.5mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 0.4mm, as drawn, 99.9+%