Merck
  • A systematic evaluation of agglomeration of Ag and TiO2 nanoparticles under freshwater relevant conditions.
A systematic evaluation of agglomeration of Ag and TiO2 nanoparticles under freshwater relevant conditions.
Environmental pollution (Barking, Essex : 1987) (2014-07-08)
Emel Topuz, Laura Sigg, Ilhan Talinli
ABSTRACT
This study aims to investigate effects of freshwater components in order to predict agglomeration behavior of silver nanoparticles coated with citrate (AgNP-Cit), polyvinylpyrrolidone (AgNP-PVP), and of TiO2 nanoparticles. Agglomeration studies were conducted in various media based on combinations of ions, natural organic matter (humic, fulvic acid) and surfactants (sodium dodecyl sulfate, alkyl ethoxylate), at a constant ionic strength of 10 mM over time for up to 1 week. Agglomeration level of AgNP-Cit and TiO2 was mostly dependent on the concentration of Ca(2+) in media, and their size strongly increased to micrometer scale over 1 week. However, AgNP-Cit and TiO2 were stabilized to particle size around 500 nm in the presence of NOM, surfactants and carbonate over 1 week. AgNP-PVP maintained their original size in all media except in the presence of Mg(2+) ions which led to significant agglomeration. Behavior of these engineered nanoparticles was similar in a natural freshwater medium.
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
Silver nanowires, diam. × L 100 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Kollidon® 25
Titanium, IRMM®, certified reference material, 0.5 mm foil
Sigma-Aldrich
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Silver nanowires, diam. × L 50 nm × 6 μm, 0.5% (isopropyl alcohol suspension)
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Titanium, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, wire, diam. 1.0 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 2.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Silver, powder, <250 μm, 99.99% trace metals basis
Sigma-Aldrich
Silver, wool, diam. 0.05 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Titanium, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Sigma-Aldrich
Silver, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 0.1 mm, 99.9% trace metals basis
Sigma-Aldrich
Silver, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Povidone, meets USP testing specifications
Sigma-Aldrich
Silver, conductive paste