Merck
  • Characterization of polychlorinated dibenzo-p-dioxins and dibenzofurans, dioxin-like polychlorinated biphenyls, and polychlorinated naphthalenes in the environment surrounding secondary copper and aluminum metallurgical facilities in China.
Characterization of polychlorinated dibenzo-p-dioxins and dibenzofurans, dioxin-like polychlorinated biphenyls, and polychlorinated naphthalenes in the environment surrounding secondary copper and aluminum metallurgical facilities in China.
Environmental pollution (Barking, Essex : 1987) (2014-07-02)
Jicheng Hu, Minghui Zheng, Wenbin Liu, Zhiqiang Nie, Changliang Li, Guorui Liu, Ke Xiao
ABSTRACT
Unintentionally produced persistent organic pollutants (UP-POPs) were determined in ambient air from around five secondary non-ferrous metal processing plants in China, to investigate the potential impacts of the emissions of these plants on their surrounding environments. The target compounds were polychlorinated dibenzo-p-dioxins and dibenzofurans (PCDD/Fs), dioxin-like polychlorinated biphenyls (dl-PCBs), and polychlorinated naphthalenes (PCNs). The PCDD/F, dl-PCB, and PCN concentrations in the ambient air downwind of the plants were 4.70-178, 8.23-7520 and 152-4190 pg/m(3), respectively, and the concentrations upwind of the plants were lower. Clear correlations were found between ambient air and stack gas concentrations of the PCDD/Fs, dl-PCBs, and PCNs among the five plants, respectively. Furthermore, the UP-POPs homolog and congener patterns in the ambient air were similar to the patterns in the stack gas samples. These results indicate that UP-POPs emissions from the plants investigated have obvious impacts on the environments surrounding the plants.
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Dibenzo[b,d]furan, BCR®, certified reference material
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Sigma-Aldrich
Dibenzofuran, 98%
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Copper, insulated wire, 20m, conductor diameter 1.0mm, insulation thickness 0.25mm, PTFE (polytetrafluoroethylene) insulation
Copper - O.F.H.C., foil, 300x300mm, thickness 0.1mm, half hard, 99.95+%
Copper, insulated wire, 1m, conductor diameter 0.45mm, insulation thickness 0.25mm, PTFE (polytetrafluoroethylene) insulation
Copper - O.F.H.C., tube, 500mm, outside diameter 10mm, inside diameter 8mm, wall thickness 1.0mm, 99.95+%
Aluminum, wire reel, 1000m, diameter 0.30mm, as drawn, 99.5%
Copper - O.F.H.C., foil, 150x150mm, thickness 0.25mm, annealed, 99.95+%
Copper - O.F.H.C., foil, 0.5m coil, thickness 0.05mm, coil width 300mm, half hard, 99.95+%