Merck
  • Clinical significance of leukotriene b4 and extracellular matrix metalloproteinase inducer in acute coronary syndrome.
Clinical significance of leukotriene b4 and extracellular matrix metalloproteinase inducer in acute coronary syndrome.
Clinical and investigative medicine. Medecine clinique et experimentale (2013-12-07)
Shasha Xu, Lijiang Tang, Yafei Mi, Jianjun Jiang, Min Zhu, Baoguo Chen, Bin Wang, Tao Li, Chenkai Xu, Jiaochen Wang, Xianben Lu, Weili Ge, Lin Wang, Jie Huang
ABSTRACT
Leukotriene B4 (LTB4) and extracellular matrix metalloproteinase (EMMPRIN) have been suggested as modulators of atherosclerotic plaque instability. This study sought to evaluate the potential diagnostic implication of LTB4 and EMMPRIN in patients with acute coronary syndrome (ACS). Patients (n=153) who underwent coronary angiography, including 105 patients diagnosed with ACS, were divided into four groups: stable angina pectoris (SAP, n=19), unstable angina pectoris (UAP, n=39), acute myocardial infarction (AMI, n=66) and control (with normal coronary angiography, n=29). EMMPRIN expression in peripheral blood mononuclear cells was determined by flow cytometry and serum LTB4 levels were measured by ELISA. To examine whether LTB4 can regulate the expression of EMMPRIN and matrix metalloproteinases (MMPs) in macrophages, differentiated THP-1 macrophages were stimulated with different concentrations of LTB4 (10-10-10-7mmol/L). Expression of EMMPRIN was evaluated by Western blotting. MMP-9 mRNA expression and enzymatic activity were determined by RT-PCR and SDS-PAGE gelatin zymography. Serum LTB4 concentration was significantly higher in AMI and UAP groups, compared with control and SAP groups (p < 0.01). Subgroups analysis showed that LTB4 was significantly higher in the AMI < 24h group, compared with the AMI > 24h group. Expression of EMMPRIN on circulating monocytes was significantly higher in patients with UAP and AMI (> 24h), compared with control, SAP and AMI (< 24h) groups (p < 0.05). In vitro study showed LTB4 up-regulated the expression of EMMPRIN, as well as the expression and activity of MMP-9, in cultured THP-1-derived macrophages (p < 0.05). LTB4 and EMMPRIN are associated with the pathogenesis of ACS and may be potential biomarkers for patients with ACS.