Merck
  • Impact of cytochrome P450 3A5 polymorphism in graft livers on the frequency of acute cellular rejection in living-donor liver transplantation.
Impact of cytochrome P450 3A5 polymorphism in graft livers on the frequency of acute cellular rejection in living-donor liver transplantation.
Pharmacogenetics and genomics (2014-06-10)
Miwa Uesugi, Mio Kikuchi, Haruka Shinke, Tomohiro Omura, Atsushi Yonezawa, Kazuo Matsubara, Yasuhiro Fujimoto, Shinya Okamoto, Toshimi Kaido, Shinji Uemoto, Satohiro Masuda
ABSTRACT
We investigated whether the cytochrome P450 3A5*3 (CYP3A5*3) genotype affects tacrolimus pharmacokinetics and the risk of acute cellular rejection in living-donor liver transplant patients in Japan. Between July 2004 and June 2011, we enrolled 410 living-donor liver transplant patients receiving tacrolimus. Biopsy specimens of intestinal mucosa and graft liver at surgery were obtained to examine the mRNA expression of CYP3A subfamilies as well as the genotyping of CYP3A5*3 polymorphism. The CYP3A5 genotype in the native intestine had no significant effect on the occurrence of acute cellular rejection between postoperative days 14 and 23 in cases with identical or compatible ABO blood types (11.5% for the CYP3A5*1 allele vs. 7.4% for CYP3A5*3/*3; P=0.2643), although the concentration/dose ratio of tacrolimus was significantly higher in patients with the intestinal CYP3A5*3/*3 genotype than in those with the CYP3A5*1 allele for 5 post-transplant weeks. However, patients who received a graft liver with the CYP3A5*1 allele showed a higher rate of acute cellular rejection than those who received a graft liver with the CYP3A5*3/*3 genotype (14.5 vs. 5.7%; P=0.0134). The relative risk for acute cellular rejection associated with the CYP3A5*1 liver allele was 2.629 (P=0.018, Cox regression model). Consequently, graft liver CYP3A5*1 genotype might increase the risk for acute cellular rejection after living-donor liver transplantation, possibly by associating with the local hepatic tacrolimus concentration. The target level of tacrolimus may be affected by the CYP3A5*3 genotype of the liver, rather than by that of the small intestine, after postoperative day 14.
Sigma-Aldrich
FK-506 monohydrate, ≥98% (HPLC)