Merck
  • Growth, serum biochemistry, complement activity, and liver gene expression responses of Pekin ducklings to graded levels of cultured aflatoxin B1.
Growth, serum biochemistry, complement activity, and liver gene expression responses of Pekin ducklings to graded levels of cultured aflatoxin B1.
Poultry science (2014-06-07)
X Chen, N Horn, P F Cotter, T J Applegate
ABSTRACT
A 14-d study was conducted to evaluate the effects of cultured aflatoxin B1 (AFB1) on performance, serum biochemistry, serum natural antibody and complement activity, and hepatic gene expression parameters in Pekin ducklings. A total of 144 male Pekin ducklings were weighed, tagged, and randomly allotted to 4 dietary treatments containing 4 concentrations of AFB1 (0, 0.11, 0.14, and 0.21 mg/kg) from 0 to 14 d of age (6 cages per diet; 6 ducklings per cage). Compared with the control group, there was a 10.9, 31.7, and 47.4% (P < 0.05) decrease in cumulative BW gain with 0.11, 0.14, and 0.21 mg of AFB1/kg of diet, respectively, but feed efficiency was not affected. Increasing concentrations of AFB1 reduced cumulative BW gain and feed intake both linearly and quadratically, and regression equations were developed with r(2) ≥0.73. Feeding 0.11 to 0.21 mg of AFB1/kg reduced serum glucose, creatinine, albumin, total protein, globulin, Ca, P, and creatine phosphokinase linearly, whereas serum urea N, Cl, alkaline phosphatase, and aspartate amino transferase concentrations increased linearly with increasing AFB1 (P < 0.05). Additionally, 0.11 to 0.21 mg of AFB1/kg diets impaired classical and alternative complement pathways in the duckling serum when tested by lysis of rabbit, human type O, and horse erythrocytes, and decreased rabbit and horse agglutinins (P < 0.05). Liver peroxisome proliferator activated receptor α (PPARα) expression was linearly downregulated by AFB1 (P < 0.01). Results from this study indicate that for every 0.10 mg/kg increase in dietary AFB1, cumulative feed intake and BW gain decrease approximately 230 and 169 g per duckling from hatch to 14 d; and that AFB1 at very low concentrations can significantly impair liver function and gene expression, and innate immune dynamics in Pekin ducklings.
Product Description
Aflatoxin B1 solution, 3.79 μg/g in acetonitrile, ERM®, certified reference material
Supelco
Aflatoxin B1 solution, 3 μg/mL in benzene:acetonitrile (98:2), analytical standard