Merck
  • Vildagliptin compared to glimepiride on post-prandial lipemia and on insulin resistance in type 2 diabetic patients.
Vildagliptin compared to glimepiride on post-prandial lipemia and on insulin resistance in type 2 diabetic patients.
Metabolism: clinical and experimental (2014-05-31)
Giuseppe Derosa, Aldo Bonaventura, Lucio Bianchi, Davide Romano, Elena Fogari, Angela D'Angelo, Pamela Maffioli
ABSTRACT
To evaluate the effects of vildagliptin compared to glimepiride on glycemic control, insulin resistance and post-prandial lipemia. 167 type 2 diabetic patients, not adequately controlled by metformin, were randomized to vildagliptin 50 mg twice a day or glimepiride 2 mg three times a day for 6 months, in a double blind, randomized clinical trial. We evaluated: body mass index (BMI), glycemic control, fasting plasma insulin (FPI), homeostasis model assessment insulin resistance index (HOMA-IR), fasting plasma proinsulin (FPPr), glucagon, lipid profile, resistin, retinol binding protein-4 (RBP-4), visfatin and vaspin. Furthermore, at the randomization and at the end of the study all patients underwent an euglycemic hyperinsulinemic clamp to evaluate M value and an oral fat load. Despite a similar decrease of glycated hemoglobin, there were an increase of body weight with glimepiride + metformin and a decrease with vildagliptin + metformin. Fasting plasma insulin increased with glimepiride + metformin, while it did not change with vildagliptin + metformin. Vildagliptin + metformin improved lipid profile. Regarding insulin sensitivity, vildagliptin + metformin increased M value. Resistin, RBP-4, vaspin and visfatin were decreased by vildagliptin + metformin, but in group to group comparison, only vaspin reduction resulted statistically significant. Vildagliptin + metformin reduced post-prandial lipemia and insulinemia compared to glimepiride + metformin. Vildagliptin, in addition to metformin, was more effective than glimepiride + metformin in reducing insulin resistance and post-prandial lipemia.
Sigma-Aldrich
Adamantane, ≥99%
Sigma-Aldrich
Glimepiride, ≥98% (HPLC), solid
Glimepiride, European Pharmacopoeia (EP) Reference Standard
Glimepiride for system suitability, European Pharmacopoeia (EP) Reference Standard