Merck
  • Acylated phenylethanoid glycosides, echinacoside and acteoside from Cistanche tubulosa, improve glucose tolerance in mice.
Acylated phenylethanoid glycosides, echinacoside and acteoside from Cistanche tubulosa, improve glucose tolerance in mice.
Journal of natural medicines (2014-04-22)
Toshio Morikawa, Kiyofumi Ninomiya, Mio Imamura, Junji Akaki, Shota Fujikura, Yingni Pan, Dan Yuan, Masayuki Yoshikawa, Xiaoguang Jia, Zheng Li, Osamu Muraoka
ABSTRACT
Acylated phenylethanoid glycosides, echinacoside (1) and acteoside (2), principal constituents in stems of Cistanche tubulosa (Orobanchaceae), inhibited the increase in postprandial blood glucose levels in starch-loaded mice at doses of 250-500 mg/kg p.o. These compounds (1 and 2) also significantly improved glucose tolerance in starch-loaded mice after 2 weeks of continuous administration at doses of 125 and/or 250 mg/kg/day p.o. without producing significant changes in body weight or food intake. In addition, several constituents from C. tubulosa, including 1 (IC50 = 3.1 μM), 2 (1.2 μM), isoacteoside (3, 4.6 μM), 2'-acetylacteoside (4, 0.071 μM), tubulosides A (5, 8.8 μM) and B (9, 4.0 μM), syringalide A 3-O-α-L-rhamnopyranoside (10, 1.1 μM), campneoside I (13, 0.53 μM), and kankanoside J1 (14, 9.3 μM), demonstrated potent rat lens aldose reductase inhibitory activity. In particular, the potency of compound 4 was similar to that of epalrestat (0.072 μM), a clinical aldose reductase inhibitor.
Sigma-Aldrich
Verbascoside, ≥99% (HPLC)
Sigma-Aldrich
Echinacoside, 98% (HPLC)
Supelco
Echinacoside, analytical standard
Verbascoside, primary reference standard