Merck
  • Phthalate and bisphenol A exposure among pregnant women in Canada--results from the MIREC study.
Phthalate and bisphenol A exposure among pregnant women in Canada--results from the MIREC study.
Environment international (2014-04-09)
Tye E Arbuckle, Karelyn Davis, Leonora Marro, Mandy Fisher, Melissa Legrand, Alain LeBlanc, Eric Gaudreau, Warren G Foster, Voleak Choeurng, William D Fraser
ABSTRACT
Bisphenol A (BPA) and phthalates are endocrine disruptors possibly linked to adverse reproductive and neurodevelopmental outcomes. These chemicals have commonly been measured in urine in population surveys; however, such data are limited for large populations of pregnant women, especially for the critical first trimester of pregnancy. The aim of the study was to measure BPA and phthalate metabolites in first trimester urine samples collected in a large national-scale pregnancy cohort study and to identify major predictors of exposure. Approximately 2000 women were recruited in the first trimester of pregnancy from ten sites across Canada. A questionnaire was administered to obtain demographic and socio-economic data on participants and a spot urine sample was collected and analyzed for total BPA (GC-MS/MS) and 11 phthalate metabolites (LC-MS/MS). The geometric mean (GM) maternal urinary concentration of total BPA, uncorrected for specific gravity, was 0.80 (95% CI 0.76-0.85) μg/L. Almost 88% of the women had detectable urinary concentrations of BPA. An analysis of urinary concentrations of BPA by maternal characteristics with specific gravity as a covariate in the linear model showed that the geometric mean concentrations: (1) decreased with increasing maternal age, (2) were higher in current smokers or women who quit during pregnancy compared to never smokers, and (3) tended to be higher in women who provided a fasting urine sample and who were born in Canada, and had lower incomes and education. Several of the phthalate metabolites analyzed were not prevalent in this population (MCHP, MMP, MiNP, MOP), with percentages detectable at less than 15%. The phthalate metabolites with the highest measured concentrations were MEP (GM: 32.02 μg/L) and MnBP (GM: 11.59 μg/L). MBzP urinary concentrations decreased with maternal age but did not differ by time of urine collection; whereas the DEHP metabolites tended to be higher in older women and when the urine was collected later in the day. This study provides the first biomonitoring results for the largest population of pregnant women sampled in the first trimester of pregnancy. The results indicate that exposure among this population of pregnant women to these chemicals is comparable to or even lower than that observed in a Canadian national population-based survey.
Product Description
Phthalic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Potassium hydrogen phthalate, BioXtra, ≥99.95%
Sigma-Aldrich
Phthalic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Phthalic acid, reagent grade, 98%
Sigma-Aldrich
Phthalic acid, puriss. p.a., ≥99.5% (T)
Supelco
Phthalic acid, analytical standard
Supelco
Potassium phthalate monobasic, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Bisphenol A, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Potassium phthalate monobasic, puriss. p.a., ≥99.5% (T)
Sigma-Aldrich
Potassium phthalate monobasic, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Bisphenol A, 97%
Sigma-Aldrich
Phthalic acid dipotassium salt, 98%
Sigma-Aldrich
Potassium phthalate monobasic, ACS reagent, acidimetric standard
Supelco
Bisphenol A, ≥99%
Supelco
Potassium phthalate monobasic, reference material for titrimetry, certified by BAM, >99.5%