Merck
  • The impact of R-VACOP-B and interim FDG-PET/CT on outcome in primary mediastinal large B cell lymphoma.
The impact of R-VACOP-B and interim FDG-PET/CT on outcome in primary mediastinal large B cell lymphoma.
Annals of hematology (2014-03-07)
Abraham Avigdor, Tsvi Sirotkin, Meirav Kedmi, Elena Ribakovsy, Miriam Berkowicz, Yaron Davidovitz, Abraham Kneller, Drorit Merkel, Yulia Volchek, Tima Davidson, Elinor Goshen, Sara Apter, Avichai Shimoni, Isaac Ben-Bassat, Arnon Nagler
ABSTRACT
The choice of a rituximab-based regimen and the prognostic significance of interim 2-[fluorine-18]fluoro-2-deoxy-D-glucose-positron emission tomography/computed tomography (FDG-PET/CT) in primary mediastinal large B cell lymphoma (PMBCL) are debatable. We evaluated the clinical features and outcomes of 95 consecutive patients with PMBCL who were treated between 1985 and 2009. Forty-three patients received rituximab-based chemotherapy, R-VACOP-B (N = 30) or R-CHOP21 (N = 13), whereas 52 patients were treated with VACOP-B (N = 47) or CHOP21 (N = 5). Radiotherapy was not given. Patients who received rituximab had a 5-year progression-free survival (PFS) of 79 % and overall survival (OS) of 97 % compared with 58 % (p = 0.06) and 88 % (p = 0.2), respectively, without rituximab. Five-year PFS in patients treated with R-VACOP-B, R-CHOP21, VACOP-B, and CHOP21 were 83, 69, 62, and 20 %, respectively (p = 0.039). However, direct comparison showed that the difference between PFS rates in patients receiving R-VACOP-B compared to R-CHOP21 was not statistically significant (p = 0.3). None of the standard clinical risk factors predicted for PFS and OS in patients receiving rituximab (R)-chemotherapy. Mid-interim FDG-PET/CT scans were performed in 30/43 patients who received R-chemotherapy. The negative predictive values of mid-PET activity were high (100 % for R-VACOP-B and 86 % for R-CHOP21) while the positive predictive values (PPV) were relatively low (30 and 75 %, respectively). Despite the low PPV, the 5-year PFS for mid-PET-negative patients (N = 16) was significantly higher (94 %) than that for mid-PET-positive (N = 14) patients (57 %, p = 0.015). This retrospective analysis demonstrates that the superiority of VACOP-B over CHOP21 for treatment of PMBCL disappeared once rituximab was added. The potential benefit of using interim PET activity as a guide for continuing therapy in patients with PMBCL remains unclear due to the relatively low PPV.
Product Description
Vincristine sulfate, European Pharmacopoeia (EP) Reference Standard
Bleomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Etoposide for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Prednisone, ≥98%
Sigma-Aldrich
Vincristine sulfate, meets USP testing specifications
Supelco
Prednisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, BioXtra, crystalline
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, for fluorescence, mixture of bleomycin sulfate salts, lyophilized, powder or crystals, white to off-white
Sigma-Aldrich
Vincristine sulfate salt, 95.0-105.0% (HPLC), powder or crystals
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, 1.5-2.0 units/mg solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Bleomycin sulfate from Streptomyces verticillus, crystalline, 1.5-2.0 U/mg
Etoposide, European Pharmacopoeia (EP) Reference Standard