Merck
Management of acute limb ischemia in the pediatric population.
Journal of vascular surgery (2014-03-25)
Ahmed Kayssi, Furqan Shaikh, Graham Roche-Nagle, Leonardo R Brandao, Suzan A Williams, Barry B Rubin
ABSTRACT
Acute limb ischemia (ALI) in pediatric patients is rare but may lead to limb loss and life-long complications. This study reviewed the experience of a Canadian tertiary pediatric center with the medical and operative management of ALI. The medical records of inpatients diagnosed with ALI of the upper or lower limb between 1999 and 2012 were reviewed. Patient demographics, arterial clot site and etiology, intervention, anticoagulation type and duration, and short-term and long-term complications were analyzed. A total of 151 patients (45% female) presented with signs of limb ischemia, of whom 38% were aged <30 days, 46% were between 1 and 12 months, and 16% were between 1 and 18 years. Ninety-four percent of those injuries involved the lower limbs. Ninety-one percent were due to vessel catheterization, 5% were idiopathic, 1% were congenital, and 4% traumatic. Ninety-four percent were managed nonoperatively. Patients were treated with a combination of thrombolysis, unfractionated or low-molecular-weight heparin, aspirin or warfarin, or both (duration, 1 day-13 years). All patients were monitored after discharge at our institution or at their referring hospital (average, 3.4 ± 2.8 years). Fifteen percent had complications related to ALI or anticoagulation (most commonly limb length or thigh circumference discrepancy, or intracranial hemorrhage). Nineteen percent of patients died of unrelated causes (sepsis, multiorgan dysfunction, or cardiac failure). In contrast with adults, ALI in children can generally be managed nonoperatively with anticoagulation, likely because of their greater ability to develop arterial collaterals. Long-term follow-up by a multidisciplinary team of pediatric and surgical specialists and allied health professionals is integral to achieving a successful outcome in children with ALI.
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Pricing and availability is not currently available.
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Aspirin, meets USP testing specifications
Pricing and availability is not currently available.
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.