Merck
  • Electrochemical degradation of the dimethyl phthalate ester on a fluoride-doped Ti/β-PbO2 anode.
Electrochemical degradation of the dimethyl phthalate ester on a fluoride-doped Ti/β-PbO2 anode.
Chemosphere (2014-03-13)
Fernanda L Souza, José M Aquino, Kallyni Irikura, Douglas W Miwa, Manuel A Rodrigo, Artur J Motheo
ABSTRACT
The electrooxidation of the dimethyl phthalate (DMP) ester was galvanostatically carried out in a filter-press reactor using a fluoride-doped lead dioxide (β-PbO2,F) film electrodeposited on a Ti substrate. The variables investigated were the nature of the supporting electrolyte (NaCl and Na2SO4), pH (3, 7, and 10), current density (10, 20, 40, 60, and 80mAcm(-2)), and temperature (10, 20, 30, 40, and 50°C). The removal of DMP was monitored through high performance liquid chromatography (HPLC) and total organic carbon (TOC) analysis. The best conditions were obtained using Na2SO4 and at low current densities, independent of the solution pH or temperature. These conditions led to the highest levels of current efficiencies and complete combustion. However, the TOC removal levels were low, due to the generation of highly oxidized intermediates, which was confirmed by the intermediates detected by HPLC.
Sigma-Aldrich
Sodium sulfate decahydrate, BioUltra, ≥99.0% (calc. on dry substance, T)
Sigma-Aldrich
Sodium bisulfate monohydrate, ReagentPlus®, 99%
Sigma-Aldrich
Lead(II) oxide, 99.999% trace metals basis
Sigma-Aldrich
Lead(II) oxide, powder, <10 μm, ReagentPlus®, ≥99.9% trace metals basis
Sigma-Aldrich
Lead(II) oxide, puriss. p.a., ≥99.0% (KT), yellow
Sigma-Aldrich
Lead(II) oxide, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium sulfate decahydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium sulfate decahydrate, puriss. p.a., crystallized, ≥99.0% (calc. based on dry substance, T)
Sigma-Aldrich
Sodium bisulfate monohydrate, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Dimethyl phthalate, ≥99%
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium hydrogen sulfate, technical grade
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Dimethyl phthalate, ≥99%
Supelco
Dimethyl phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)