Merck
  • IL-9 regulates allergen-specific Th1 responses in allergic contact dermatitis.
IL-9 regulates allergen-specific Th1 responses in allergic contact dermatitis.
The Journal of investigative dermatology (2014-02-04)
Juan Liu, Erin Harberts, Antonella Tammaro, Nicholas Girardi, Renata B Filler, Rita Fishelevich, Angela Temann, Paula Licona-Limón, Michael Girardi, Richard A Flavell, Anthony A Gaspari
ABSTRACT
The cytokine IL-9, derived primarily from T-helper 9 (Th9) lymphocytes, promotes expansion of the Th2 subset and is implicated in the mechanisms of allergic asthma. We hypothesize that IL-9 also has a role in human allergic contact dermatitis (ACD). To investigate this hypothesis, skin biopsy specimens of positive patch-test sites from non-atopic patients were assayed using quantitative PCR and immunohistochemistry. The cytokines IFN-γ, IL-4, IL-17A, IL-9, and PU.1, a Th9 associated transcription factor, were elevated when compared with paired normal skin. Immunohistochemistry on ACD skin biopsies identified PU.1+ CD3+ and PU.1+ CD4+ cells, consistent with Th9 lymphocytes, in the inflammatory infiltrate. Peripheral blood mononuclear cells from nickel-allergic patients, but not nonallergic controls, show significant IL-9 production in response to nickel. Blocking studies with mAbs to HLA-DR (but not HLA-A, -B, -C) or chloroquine significantly reduced this nickel-specific IL-9 production. In addition, blockade of IL-9 or IL-4 enhanced allergen-specific IFN-γ production. A contact hypersensitivity model using IL-9(-/-) mice shows enhanced Th1 lymphocyte immune responses, when compared with wild-type mice, consistent with our human in vitro data. This study demonstrates that IL-9, through its direct effects on Th1 and ability to promote IL-4 secretion, has a regulatory role for Th1 lymphocytes in ACD.
Sigma-Aldrich
Nickel, wire, diam. 0.25 mm, ≥99.9%
Sigma-Aldrich
Nickel, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2400, slurry, in H2O, active catalyst
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2800, slurry, in H2O, active catalyst
Sigma-Aldrich
Nickel, powder, <1 μm, 99.8% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.25 mm, 99.995% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.125 mm, ≥99.9%
Nickel, wire reel, 200m, diameter 0.125mm, hard, 99.98%
Nickel, foil, 1m coil, thickness 0.20mm, coil width 1mm, annealed, 99%
Nickel, foil, 1m coil, thickness 0.025mm, coil width 600mm, as rolled, 99%
Nickel, tube, 200mm, outside diameter 1.3mm, inside diameter 1.1mm, wall thickness 0.1mm, hard, 99.5%
Nickel, tube, 100mm, outside diameter 3.6mm, inside diameter 3.5mm, wall thickness 0.05mm, hard, 99.5%
Nickel, foil, 25mm disks, thickness 0.075mm, annealed, 99%
Nickel, tube, 100mm, outside diameter 5.5mm, inside diameter 5mm, wall thickness 0.25mm, hard, 99.5%
Nickel, foil, 25mm disks, thickness 0.007mm, 99.95%
Nickel, foil, 25mm disks, thickness 0.025mm, 99.9%
Nickel, tube, 3mm, outside diameter 1.5mm, inside diameter 1.3mm, wall thickness 0.1mm, hard, 99.5%
Nickel, foil, 25mm disks, thickness 0.075mm, as rolled, 99.98%
Nickel, foil, 20m coil, thickness 0.05mm, coil width 2mm, annealed, 99%
Nickel, foil, 2m coil, thickness 0.125mm, annealed, 99%
Nickel, foil, 25x25mm, thickness 1.0mm, as rolled, 99.99+%
Nickel, foil, 10mm disks, thickness 0.0025mm, 99.95%
Nickel, foil, 150x150mm, thickness 0.25mm, annealed, 99.98%
Nickel, wire reel, 100m, diameter 0.5mm, annealed, 99%
Nickel, foil, 0.5m coil, thickness 0.015mm, 99.9%
Nickel, wire reel, 0.2m, diameter 1.0mm, as drawn, 99.99+%
Nickel, foil, 4mm disks, thickness 0.015mm, 99.9%
Nickel, foil, 2m coil, thickness 0.30mm, as rolled, 99.98%
Nickel, foil, not light tested, 50x50mm, thickness 0.002mm, 99.95%