Merck
  • Synthesis and in vitro biological evaluation of 2,6,9-trisubstituted purines targeting multiple cyclin-dependent kinases.
Synthesis and in vitro biological evaluation of 2,6,9-trisubstituted purines targeting multiple cyclin-dependent kinases.
European journal of medicinal chemistry (2012-07-10)
Marek Zatloukal, Radek Jorda, Tomáš Gucký, Eva Řezníčková, Jiří Voller, Tomáš Pospíšil, Veronika Malínková, Helena Adamcová, Vladimír Kryštof, Miroslav Strnad
ABSTRACT
Several inhibitors of cyclin-dependent kinases (CDKs), including the 2,6,9-trisubstituted purine derivative roscovitine, are currently being evaluated in clinical trials as potential anticancer drugs. Here, we describe a new series of roscovitine derivatives that show increased potency in vitro. The series was tested for cytotoxicity against six cancer cell lines and for inhibition of CDKs. For series bearing 2-(hydroxyalkylamino) moiety, cytotoxic potency strongly correlated with anti-CDK2 activity. Importantly, structural changes that increase biochemical and anticancer activities of these compounds also increase elimination half-life. The most potent compounds were investigated further to assess their ability to influence cell cycle progression, p53-regulated transcription and apoptosis. All the observed biological effects were consistent with inhibition of CDKs involved in the regulation of cell cycle and transcription.
Sigma-Aldrich
3-Methoxybenzylamine, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Methoxybenzylamine, 98%
Pricing and availability is not currently available.