Merck
Improving cell-free protein synthesis for stable-isotope labeling.
Journal of biomolecular NMR (2007-01-24)
Takayoshi Matsuda, Seizo Koshiba, Naoya Tochio, Eiko Seki, Noriyuki Iwasaki, Takashi Yabuki, Makoto Inoue, Shigeyuki Yokoyama, Takanori Kigawa
ABSTRACT
Cell-free protein synthesis is suitable for stable-isotope labeling of proteins for NMR analysis. The Escherichia coli cell-free system containing potassium acetate for efficient translation (KOAc system) is usually used for stable-isotope labeling, although it is less productive than other systems. A system containing a high concentration of potassium L-glutamate (L-Glu system), instead of potassium acetate, is highly productive, but cannot be used for stable-isotope labeling of Glu residues. In this study, we have developed a new cell-free system that uses potassium D-glutamate (D-Glu system). The productivity of the D-Glu system is approximately twice that of the KOAc system. The cross peak intensities in the 1H-15N HSQC spectrum of the uniformly stable-isotope labeled Ras protein, prepared with the D-Glu system, were similar to those obtained with the KOAc system, except that the Asp intensities were much higher for the protein produced with the D-Glu system. These results indicate that the D-Glu system is a highly productive cell-free system that is especially useful for stable-isotope labeling of proteins.
Sigma-Aldrich
In Vitro Protein Expression (iPE) Kit II
Pricing and availability is not currently available.