Merck
Listening in on bacteria: acyl-homoserine lactone signalling.
Nature reviews. Molecular cell biology (2002-09-05)
Clay Fuqua, E Peter Greenberg
ABSTRACT
Bacterial cell-to-cell signalling has emerged as a new area in microbiology. Individual bacterial cells communicate with each other and co-ordinate group activities. Although a lot of detail is known about the mechanisms of a few well-characterized bacterial communication systems, other systems have been discovered only recently. Bacterial intercellular communication has become a target for the development of new anti-virulence drugs.
Sigma-Aldrich
N-(3-Hydroxydodecanoyl)-DL-homoserine lactone, ≥97% (HPLC)
Sigma-Aldrich
N-(3-Hydroxytetradecanoyl)-DL-homoserine lactone, ≥96% (HPLC), carbon 64.7-67.3 %
Sigma-Aldrich
N-Hexanoyl-L-homoserine lactone, ≥96% (HPLC)
Sigma-Aldrich
N-Octanoyl-L-homoserine lactone, ≥96% (HPLC)
Sigma-Aldrich
N-Decanoyl-L-homoserine lactone, ≥96% (HPLC)
Sigma-Aldrich
N-(3-Hydroxyoctanoyl)-DL-homoserine lactone, ≥95% (sum of isomers, HPLC)
Sigma-Aldrich
N-(3-Hydroxydecanoyl)-DL-homoserine lactone
Sigma-Aldrich
N-Heptanoyl-L-homoserine lactone, ≥96% (HPLC)
Sigma-Aldrich
N-Dodecanoyl-L-homoserine lactone, ≥96% (HPLC)