Merck
  • [The use of benzyl alcohol and amyl-m-cresol (Strepsils) in the oral cavity. (Review of the literature and first clinical experiences)].
[The use of benzyl alcohol and amyl-m-cresol (Strepsils) in the oral cavity. (Review of the literature and first clinical experiences)].
Fogorvosi szemle (1998-06-11)
L Gáspár, J Turi, B Z Tóth, C Suri, P Vágó
ABSTRACT
The authors summarised the literature data of benzylalcohol and amyl-m-cresol. In the clinical practice they used Strepsils in the treatment of 22 patients suffered from oral inflammatic diseases and in the 20 prophylactic cases of oral surgical interventions. The preliminary clinical report shows that, the Strepsils seems to be very effective in the treatment of oral inflammations and in the prophylaxis too.
Product Description
Supelco
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzyl alcohol, analytical standard
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG
Supelco
Benzyl alcohol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Benzyl alcohol, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzyl alcohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%
Sigma-Aldrich
Benzyl alcohol, puriss., meets analytical specification of Ph. Eur., BP, NF, 99-100.5% (GC)
Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Benzyl alcohol, USP, 98.0-100.5%