Merck
  • Oral vitamin D or calcium carbonate in the prevention of renal bone disease?
Oral vitamin D or calcium carbonate in the prevention of renal bone disease?
Current opinion in nephrology and hypertension (1996-07-01)
A Argilés, G Mourad, C Mion
ABSTRACT
It is well known that hyperparathyroidism begins early in renal failure and progresses, probably not linearly, throughout the natural course of renal diseases and dialysis therapy. Recent progress in basic medical science has improved our understanding of the mechanisms by which the classically known stimuli for parathyroid hormone synthesis and secretion may act, including hypocalcaemia, hyperphosphataemia and vitamin D3 metabolism disturbances. In the treatment of hyperparathyroidism, although some authors stress the benefit of treating one of these stimuli, it is probably more effective to combine the treatment of them all. There is conclusive recent work showing the efficacy of using both CaCO3 and vitamin D3, either in chronic renal failure or in dialysis patients at every stage of hyperparathyroidism. Therefore, the treatment of hyperparathyroidism should start early, long before dialysis, and it should aim to correct any of the causal factors. Both CaCO3 and vitamin D3 derivatives may be used in the prevention and treatment of renal bone disease. The limits of this association are the increasingly often reported adynamic bone disease, which in our experience has not yet given major clinical problems, and hyperphosphataemia. Uncontrolled serum phosphate levels would counterbalance the beneficial effect of vitamin D3 derivatives on hyperparathyroidism.
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Supelco
Calcium carbonate, reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Calcium carbonate, ACS reagent, chelometric standard, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium carbonate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Supelco
Calcium carbonate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Calcium carbonate, ReagentPlus®
Sigma-Aldrich
Calcium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Calcium carbonate, ACS reagent, ≥99.0%, powder
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%