Merck
  • n-hexane polyneuropathy in Japan: a review of n-hexane poisoning and its preventive measures.
n-hexane polyneuropathy in Japan: a review of n-hexane poisoning and its preventive measures.
Environmental research (1993-07-01)
Y Takeuchi
ABSTRACT
n-Hexane is used in industry as a solvent for adhesive, dry cleaning, and vegetable oil extraction. In 1963, the first case of severe polyneuropathy suspected to be caused by n-hexane was referred to us. Case studies, animal experiments, and field surveys on n-hexane poisoning were conducted, and preventive measures like threshold limit value revision and biological monitoring were also studied. I review a brief history of our investigations on n-hexane poisoning and its preventive measures in Japan. n-Hexane could cause overt polyneuropathy in workers exposed to more than 100 ppm time-weighted average concentrations [TWA]. The present threshold limit value of 40 ppm in Japan is considered low enough to prevent subclinical impairment of peripheral nerve caused by n-hexane. Urinary 2,5-hexanedione could be a good indicator for biological monitoring of n-hexane exposure. About 2.2 mg/liter of 2,5-hexanedione measured by our improved method corresponds to exposure of 40 ppm (TWA) of n-hexane.
Product Description
Supelco
Hexane, analytical standard
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)