- Beclomethasone dipropionate in the management of rhinitis--a review.
Beclomethasone dipropionate in the management of rhinitis--a review.
Annals of allergy (1982-09-01)
P Small, M Black, S Frenkiel, G N Wachs
PMID7051907
ABSTRACT
Beclomethasone dipropionate has been demonstrated to be effective in the treatment of seasonal and perennial rhinitis. Comparative studies with other medications are lacking. Adverse reactions are frequent but minor. No alterations of the hypothalamic-pituitary-adrenal axis, nasal mucosal histology or bacterial and fungal flora have been recognized.
Product Description
Beclometasone dipropionate for system suitability, European Pharmacopoeia (EP) Reference Standard
Beclometasone dipropionate for peak identification, European Pharmacopoeia (EP) Reference Standard