Merck
  • Mycophenolate mofetil vs. methotrexate for the prevention of graft-versus-host-disease--systematic review and meta-analysis.
Mycophenolate mofetil vs. methotrexate for the prevention of graft-versus-host-disease--systematic review and meta-analysis.
Leukemia research (2014-01-15)
Ron Ram, Moshe Yeshurun, Liat Vidal, Ofer Shpilberg, Anat Gafter-Gvili
ABSTRACT
We performed a systematic review and meta-analysis of all trials comparing MMF and methotrexate as GVHD prophylaxis. Our search yielded 11 studies; 3 were randomized-control trials (RCTs). While the incidence of grades 2-4 acute GVHD was comparable, the incidence of grades 3 and 4 acute GVHD was higher in patients given MMF (RR 1.61; 95% CI 1.18-2.30). Incidence of mucositis was lower (RR 0.35; 95% CI 0.25-0.49) and time to engraftment was shorter (mean difference (-3.6); 95% CI -5.5 to -1.7) in patients given MMF. All other analyzed transplantation outcomes were comparable. We conclude that MMF, compared to methotrexate, is associated with increased severity of acute GVHD. Robustness of these results is hampered by the small number of RCTs.
Sigma-Aldrich
Methotrexate hydrate, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Sigma-Aldrich
Methotrexate hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Methotrexate hydrate, powder, BioReagent, suitable for cell culture, ≥98% (HPLC)
Mycophenolate mofetil for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mycophenolate mofetil, ≥98% (HPLC)
Mycophenolate mofetil, European Pharmacopoeia (EP) Reference Standard