Merck
Therapeutic and cosmetic applications of mangiferin: a patent review.
Expert opinion on therapeutic patents (2013-09-27)
Manasi Telang, Sivakami Dhulap, Anita Mandhare, Rajkumar Hirwani
ABSTRACT
Mangiferin, a natural C-glucoside xanthone [2-C-β-D-glucopyranosyl-1, 3, 6, 7-tetrahydroxyxanthone], is abundantly present in young leaves and stem bark of the mango tree. The xanthonoid structure of mangiferin with C-glycosyl linkage and polyhydroxy components contributes to its free radical-scavenging ability, leading to a potent antioxidant effect as well as multiple biological activities. An extensive search was carried out to collect patent information on mangiferin and its derivatives using various patent databases spanning all priority years to date. The patents claiming therapeutic and cosmetic applications of mangiferin and its derivatives were analyzed in detail. The technology areas covered in this article include metabolic disorders, cosmeceuticals, multiple uses of the same compound, miscellaneous uses, infectious diseases, inflammation, cancer and autoimmune disorders, and neurological disorders. Mangiferin has the potential to modulate multiple molecular targets including nuclear factor-kappa B (NF-κB) signaling and cyclooxygenase-2 (COX-2) protein expression. Mangiferin exhibits antioxidant, antidiabetic, antihyperuricemic, antiviral, anticancer and antiinflammatory activities. The molecular structure of mangiferin fulfils the four Lipinski's requisites reported to favor high bioavailability by oral administration. There is no evidence of adverse side effects of mangiferin so far. Mangiferin could thus be a promising candidate for development of a multipotent drug.
Sigma-Aldrich
Mangiferin Mangifera indica
Supelco
Mangiferin, analytical standard