Merck
An evolving role of piperazine moieties in drug design and discovery.
Mini reviews in medicinal chemistry (2013-07-31)
Rahul V Patel, Se Won Park
ABSTRACT
This article purposes to provide insights to piperazine based molecular designs that will facilitate drug discovery program in future. In our pursuit to summarize the reservoir of bioactive agents, and in line with the synthetic economy of new heterocycles, many new roles are being identified for the multiple biotargets of piperazine moieties. We mark out how series of different scaffolds provide an extensive range of various piperazine-based analogues displaying antimicrobial, antituberculosis, anticancer, antiviral and antimalarial activities. We believe that piperazine family of compounds, and their various co-components, highlight the existence of several potential leads for the furtherance of novel bioactive agents.
Sigma-Aldrich
Piperazine, ReagentPlus®, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Piperazine, BioUltra, anhydrous, ≥99.0% (T)