Merck
Sense and nonsense of high-dose cytarabine for acute myeloid leukemia.
Blood (2013-01-05)
Bob Löwenberg
ABSTRACT
High-dose cytarabine applied during remission induction or as consolidation after attainment of a complete remission has become an established element in the treatment of adults with acute myeloid leukemia. Recent evidence has challenged the need for these exceptionally high-dose levels of cytarabine. In this review, we present a reappraisal of the usefulness of high-dose cytarabine for acute myeloid leukemia treatment.
Product Description
Cytarabine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cytosine β-D-arabinofuranoside, crystalline, ≥90% (HPLC)