Merck
  • [Research progress of the inhibitory effect of deuterium-depleted water on cancers].
[Research progress of the inhibitory effect of deuterium-depleted water on cancers].
Nan fang yi ke da xue xue bao = Journal of Southern Medical University (2012-10-19)
Hongqiang Wang, Cong Liu, Weiyi Fang, Huiling Yang
ABSTRACT
Deuterium is an important predisposing factor for cancer. Deuterium-depleted Water, also known as low deuterium water, ultra-light water or no deuterium water, can be obtained by removing deuterium from natural water. Studies have shown that water with a low deuterium concentration (<65% percent of volume) can inhibit cancer growth. Clinical trials demonstrated that drinking DDW (10-20 ppm) caused growth arrest of malignant cells in cancer patients and significantly prolonged the patient survival with also improved quality of life. A wide range of anti-cancer drugs in current use are associated with severe adverse effects, while deuterium-depleted water appears to have virtually no pharmacological side effects and is convenient to administer. The authors review the advances in the researches of anti-cancer effects and the underlying mechanisms of deuterium-depleted water.
Sigma-Aldrich
Deuterium, 99.9 atom % D