Merck
  • CNS insulin signaling in the control of energy homeostasis and glucose metabolism - from embryo to old age.
CNS insulin signaling in the control of energy homeostasis and glucose metabolism - from embryo to old age.
Trends in endocrinology and metabolism: TEM (2012-12-26)
Merly C Vogt, Jens C Brüning
ABSTRACT
Central nervous system (CNS) insulin signaling regulates energy and glucose homeostasis by acting on hypothalamic neurocircuits and higher brain circuits such as the dopaminergic system. However, overnutrition, obesity, and type 2 diabetes mellitus (T2DM) induce insulin resistance selectively in different regions of the brain, thereby impairing energy homeostasis and augmenting disease progression. Moreover, fetal hyperinsulinemia in response to maternal overnutrition, obesity, and diabetes disrupts hypothalamic neurocircuit development and predisposes to metabolic disorders later in life. In light of the current obesity and diabetes epidemic, we review the molecular basis of insulin action and resistance in the CNS, mechanisms which are causal to the development of these metabolic disorders, both in the neonate and in the adult.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
D-(+)-Glucose solution, 1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture