Merck
Fatal adverse effects of injected ceftriaxone sodium in China.
Pharmacoepidemiology and drug safety (2012-07-05)
Yi Yao, Ranyun Zhou, Ying Wang
ABSTRACT
The incidence of adverse reactions to ceftriaxone, which is widely used in China, has gradually increased, with an associated increase in patient fatality. An analysis of the reported data from articles published in China highlights the importance and extent of this growing public health problem. We identified previously reported cases of adverse effects to ceftriaxone sodium by searching the Chinese Medical Text Database System for reports published between January 2002 and December 2009. The references cited in the articles were examined for additional reports. A total of 17 articles were identified that cited fatal adverse reactions in 22 cases. Most patients had been treated for upper respiratory tract infection and bronchitis and had no reported history of ceftriaxone allergy. Of the patients, 77% were treated with ceftriaxone without undergoing intradermal testing, and 36% were not offered any information on intradermal testing. Concomitant intravenous medications were prescribed in 7 cases, and an overdose of ceftriaxone sodium was prescribed for two patients. The delay between drug administration and the onset of adverse drug reactions occurred within 30 min in 72.7% of the patients. Of the deaths, 73% occurred on the first day of treatment. The fatal adverse events associated with ceftriaxone occurred because of inappropriate drug usage, drug overdoses, careless medical personnel, poor medical conditions, and possibly poor drug quality. Although some deaths may be unavoidable, the risks can be minimized by the appropriate administration of ceftriaxone and further consultation with the Chinese medical profession and research.
Sigma-Aldrich
Ceftriaxone disodium salt hemi(heptahydrate), third-generation cephalosporin antibiotic
Ceftriaxone sodium, European Pharmacopoeia (EP) Reference Standard
Supelco
Ceftriaxone Sodium, Pharmaceutical Secondary Standard; Certified Reference Material