Merck
Tobramycin for the treatment of bacterial pneumonia in children.
Expert opinion on pharmacotherapy (2012-02-02)
Meenakshi Bothra, Rakesh Lodha, Sushil Kumar Kabra
ABSTRACT
Common etiological agents for community-acquired lower respiratory tract infection (LRTI) include Streptococcus pneumoniae, Hemophilus influenzae and Mycoplasma pneumoniae and can be easily managed with oral or intravenous antibiotics. However, LRTI in patients with underlying illnesses, such as cystic fibrosis (CF) and immune deficiency, or on ventilator support is difficult to manage because these are caused by Gram-negative bacilli. Tobramycin has been shown to be effective in the management of these patients. Information about the antimicrobial activity, pharmacological aspects (including pharmacokinetics and pharmacodynamics), clinical efficacy, safety and side effects of tobramycin have been covered in this review. A major advance for the use of tobramycin has occurred with its use by the inhalational route, in children with CF. The inhalation route provides the advantage of ease of administration for prolonged periods at home and allows use of very high doses. Systematic reviews suggest that tobramycin inhalation improves outcome, decreases the need for hospitalization and decreases the need for use of frequent systemic antibiotics in CF patients colonized with pseudomonas. Data on the efficacy of inhaled tobramycin in non-CF bronchiectasis are scarce, as are data on the prevention and treatment of ventilator-associated pneumonia, and on the role of combining inhaled tobramycin with systemic tobramycin. Despite limitations, this drug has the potential to be used in various conditions other than CF.
Sigma-Aldrich
Tobramycin, Aminoglycoside antibiotic
Supelco
Tobramycin, Pharmaceutical Secondary Standard; Certified Reference Material
Tobramycin for identification, European Pharmacopoeia (EP) Reference Standard