Merck
  • Gentian violet 1% solution in the treatment of wounds in the geriatric patient: a retrospective study.
Gentian violet 1% solution in the treatment of wounds in the geriatric patient: a retrospective study.
Geriatric nursing (New York, N.Y.) (2011-02-18)
Karen J Farid, Kathy Kelly, Shyla Roshin
ABSTRACT
To review the use of gentian violet 1% (GV) in a long-term care facility for the treatment of small, open wounds and extremity eschars of all sizes and thickness. The records of all the patients receiving topical GV therapy over a period of 1 year, from May 19, 2007 - May 19, 2008 were reviewed. The total sample was 70 patients (38 male, 32 female), average age 65.9 y.o., with 111 wounds (41 patients had >1 wound). The types of wounds were divided into 3 categories: 1) Split-thickness scabs (N = 37) limited to the epithelium, 2) full-thickness eschars and wounds with no depth <1 cm (N = 50), and 3) full-thickness eschars >1 cm located on the lower extremity (average 3.3 cm in diameter [N = 24]). All the wounds had been treated the same: topical application of GV to the wounds daily. None of the patients had any documented adverse events to the GV. Out of the 111 wounds, 103 healed completely. All wounds remained negative for cellulitis. 1) GV is a viable alternative topical agent for healing small, superficial wounds, ineffective scabs, and both small and large pressure ulcer eschars on lower extremities of geriatric patients, 2) there was very little evidence of scarring with the use of GV.
Sigma-Aldrich
Gentian Violet, meets USP testing specifications
Sigma-Aldrich
Crystal Violet Solution, 1%, aqueous solution
Sigma-Aldrich
Crystal Violet, indicator for the determination of the redox potential, S. No.: 785
Sigma-Aldrich
Crystal Violet, certified by the Biological Stain Commission
Sigma-Aldrich
Crystal Violet, for microscopy (Bact., Bot., Hist., Vit.), indicator (pH 0.1-2.0)
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Supelco
Crystal Violet, VETRANAL®, analytical standard
Methylrosanilinium for system suitability, European Pharmacopoeia (EP) Reference Standard
Methylrosanilinium chloride, European Pharmacopoeia (EP) Reference Standard