Merck
Erythromycin as a prokinetic agent: risk factors.
Journal of visceral surgery (2010-07-27)
S Berthet, B Charpiat, J Y Mabrut
ABSTRACT
Erythromycin (ER) is an antibiotic with prokinetic activity. This property has led to its clinical use to improve gastric emptying in patients with gastroparesis. Several papers have reported the effectiveness of ER in such patients; however few of these studies have been well-conducted methodologically with evaluation of clinical endpoints and their results are often contradictory. A benefit/risk analysis prior to prescription of ER should take into account the numerous medication interactions of this drug as well as the prevention of cardiovascular complications, most notably torsade de pointes and cardiac arrest. The aim of this paper was to provide new insights into these adverse events and allow for better-informed prescription. Risk prevention requires prescription guidelines and effective communication among surgeons, anesthesiologists and pharmacists.
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Erythromycin, for microbiological assay, European Pharmacopoeia (EP) Reference Standard