Merck
  • Application of the margin-of-exposure (MoE) approach to substances in food that are genotoxic and carcinogenic e.g.: benzo[a]pyrene and polycyclic aromatic hydrocarbons.
Application of the margin-of-exposure (MoE) approach to substances in food that are genotoxic and carcinogenic e.g.: benzo[a]pyrene and polycyclic aromatic hydrocarbons.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2009-10-13)
Diane Benford, Michael Dinovi, R Woodrow Setzer
ABSTRACT
Benzo[a]pyrene (BaP) and a number of other polycyclic aromatic hydrocarbons (PAH) are mutagenic and are also carcinogenic in rodent bioassays. Oral carcinogenicity data are not available for individual PAH other than BaP, and so BaP has been used as a marker of the carcinogenicity of, and exposure to, PAHs. Carcinogenicity studies of coal tar mixtures, considered to be representative of the genotoxic and carcinogenic PAH in food, have been used for dose-response modelling. Modelling the number of tumour-bearing mice resulted in a BMDL(10) of 0.122 mg BaP/kg-bw/day, which was lower than that for any of the individual tumours and was considered to be most appropriate since the different PAH may have different mechanisms of carcinogenicity. An average dietary exposure estimates of 0.008 microg BaP/kg-bw/day was identified from the range of national estimates. The calculated MoE was 15,000.
Product Description
Supelco
Benzo[a]pyrene solution, 100 μg/mL in cyclohexane, analytical standard
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 200 μg/mL in methylene chloride
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 1000 μg/mL in acetone
Sigma-Aldrich
Benzo[a]pyrene, ≥96% (HPLC)
Supelco
Benzo[a]pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Benzo[a]pyrene, analytical standard, for environmental analysis