Merck
  • A review of the economic evidence for acarbose in the prevention of diabetes and cardiovascular events in individuals with impaired glucose tolerance.
A review of the economic evidence for acarbose in the prevention of diabetes and cardiovascular events in individuals with impaired glucose tolerance.
International journal of clinical practice (2006-07-22)
R G Josse, A J McGuire, G B Saal
ABSTRACT
The highly prevalent, prediabetic condition of impaired glucose tolerance (IGT) confers a high risk for type 2 diabetes mellitus (T2DM) and cardiovascular disease (CVD). There is an emerging body of cost-effectiveness literature in the management of IGT. For acarbose, an alpha-glucosidase inhibitor, economic analyses have been conducted for Spain, Germany, Sweden and Canada. In Spain, acarbose was more effective and less costly (dominant) compared with placebo. In Germany, the cost per patient free of diabetes was under Pounds 800; acarbose was dominant for those at high risk for T2DM, CVD or both, and a similar outcome in the Swedish study. In Canada, acarbose was dominant compared with no intervention and very cost-effective compared with metformin [C Dollars 1798/life years gained (LYG)]. The particularly cost-effective outcomes or cost savings delivered by acarbose for IGT subjects at high risk for T2DM and/or CVD render an IGT-intervention program prioritised to such high-risk individuals an economically attractive strategy.
Product Description
Acarbose for identification, European Pharmacopoeia (EP) Reference Standard
Acarbose for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acarbose, ≥95% (HPLC)
Supelco
Acarbose, Pharmaceutical Secondary Standard; Certified Reference Material
Acarbose, European Pharmacopoeia (EP) Reference Standard