Merck
Bioconversions of ferulic acid, an hydroxycinnamic acid.
Critical reviews in microbiology (2006-08-09)
Sindhu Mathew, T Emilia Abraham
ABSTRACT
Ferulic acid is the most abundant hydroxycinnamic acid in the plant world and is ester linked to arabinose, in various plant polysaccharides such as arabinoxylans and pectins. It is a precursor to vanillin, one of the most important aromatic flavor compound used in foods, beverages, pharmaceuticals, and perfumes. This article presents an overview of the various biocatalytic routes, focusing on the relevant biotransformations of ferulic acid using plant sources, microorganisms, and enzymes.
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Supelco
trans-Ferulic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
trans-Ferulic acid, 99%
Supelco
trans-Ferulic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Ferulic acid, European Pharmacopoeia (EP) Reference Standard